missing translation for 'onlineSavingsMsg'
Learn More

Benzyl nicotinate, 98%

Catalog No. AAA1850530
Change view
Click to view available options
Quantity:
50 g
250 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1850530 250 g
AAA1850518 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1850530 Supplier Thermo Scientific Chemicals Supplier No. A1850530
Only null left

CAS: 94-44-0 | C13H11NO2 | 213.236 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 94-44-0
Molecular Formula C13H11NO2
Molecular Weight (g/mol) 213.236
MDL Number MFCD00023584
InChI Key KVYGGMBOZFWZBQ-UHFFFAOYSA-N
Synonym benzyl nicotinate, nicotinic acid benzyl ester, rubriment, pycaril, pykaryl, niacin benzyl ester, 3-pyridinecarboxylic acid, phenylmethyl ester, benzylis nicotinas, nicotinic acid, benzyl ester, nicotinsaeurebenzylester
PubChem CID 7191
ChEBI CHEBI:31268
IUPAC Name benzyl pyridine-3-carboxylate
SMILES C1=CC=C(C=C1)COC(=O)C2=CN=CC=C2

Specifications

Melting Point 21°C to 23°C
Density 1.165
Boiling Point 179°C to 180°C (5 mmHg)
Flash Point 170°C (338°F)
Refractive Index 1.57
Quantity 250 g
Beilstein 159169
Merck Index 14,6526
Formula Weight 213.24
Percent Purity 99%
Chemical Name or Material Benzyl nicotinate
Show More Show Less
Hazard Category H315-H319
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P264b-P280-P302+P352-P305+P351+P338-P332+P313-P362
EINECSNumber 202-332-3
RTECSNumber QT0850000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only