missing translation for 'onlineSavingsMsg'
Learn More

Benzyl Methyl Malonate 96.0+%, TCI America™

Catalog No. p-7132470
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M09745G 5 g
M097425G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M09745G Supplier TCI America Supplier No. M09745G
Only null left

Chemical Identifiers

CAS 52267-39-7
Molecular Formula C11H12O4
Molecular Weight (g/mol) 208.21
MDL Number MFCD00008461
InChI Key IAUZDBFOEWAQFE-UHFFFAOYSA-N
Synonym benzyl methyl malonate, 1-benzyl 3-methyl propanedioate, malonic acid 1-benzyl 3-methyl ester, propanedioic acid, methyl phenylmethyl ester, benzylmethyl malonate, methyl benzyl malonate, acmc-1b0zd, benzyl methyl propanedioate, ksc493e1f
PubChem CID 572135
IUPAC Name 1-benzyl 3-methyl propanedioate
SMILES COC(=O)CC(=O)OCC1=CC=CC=C1

Specifications

Color Yellow
Quantity 5 g
Formula Weight 208.21
Percent Purity ≥96.0% (GC)
Physical Form Liquid
Chemical Name or Material Benzyl Methyl Malonate