missing translation for 'onlineSavingsMsg'
Learn More

Benzyl methyl malonate, 95%

Catalog No. AAH5914014
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH5914014 25 g
AAH5914006 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH5914014 Supplier Thermo Scientific Chemicals Supplier No. H5914014
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 52267-39-7 | C11H12O4 | 208.21 g/mol

It is employed as a intermediate for pharmaceutical and organic synthesis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is employed as a intermediate for pharmaceutical and organic synthesis.

Solubility
Slightly soluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.

Chemical Identifiers

CAS 52267-39-7
Molecular Formula C11H12O4
Molecular Weight (g/mol) 208.21
MDL Number MFCD00008461
InChI Key IAUZDBFOEWAQFE-UHFFFAOYSA-N
Synonym benzyl methyl malonate, 1-benzyl 3-methyl propanedioate, malonic acid 1-benzyl 3-methyl ester, propanedioic acid, methyl phenylmethyl ester, benzylmethyl malonate, methyl benzyl malonate, acmc-1b0zd, benzyl methyl propanedioate, ksc493e1f
PubChem CID 572135
IUPAC Name 3-O-benzyl 1-O-methyl propanedioate
SMILES COC(=O)CC(=O)OCC1=CC=CC=C1

Specifications

Density 1.15
Boiling Point 125°C (0.5 mmHg)
Flash Point >113°C (235°F)
Refractive Index 1.502
Linear Formula C6H5CH2OCOCH2COOCH3
Quantity 25 g
Beilstein 2529986
Solubility Information Slightly soluble in water.
Formula Weight 208.21
Percent Purity 95%
Chemical Name or Material Benzyl methyl malonate
Show More Show Less
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only