missing translation for 'onlineSavingsMsg'
Learn More

Benzyl Methacrylate (stabilized with MEHQ) 98.0+%, TCI America™

Catalog No. M027925ML
Change view
Click to view available options
Quantity:
25 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M027925ML 25 mL
M0279500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M027925ML Supplier TCI America Supplier No. M027925ML

Chemical Identifiers

CAS 2495-37-6
Molecular Formula C11H12O2
Molecular Weight (g/mol) 176.215
MDL Number MFCD00053684
InChI Key AOJOEFVRHOZDFN-UHFFFAOYSA-N
Synonym benzyl methacrylate, 2-propenoic acid, 2-methyl-, phenylmethyl ester, benzylmethacrylate, methacrylic acid benzyl ester, methacrylic acid, benzyl ester, unii-z3248k2ssm, phenylmethyl 2-methylprop-2-enoate, poly benzylmethacrylate, bzma, methacrylic acid benzyl
PubChem CID 17236
IUPAC Name benzyl 2-methylprop-2-enoate
SMILES CC(=C)C(=O)OCC1=CC=CC=C1

Specifications

Color Colorless
Boiling Point 106°C
Quantity 25 mL
Formula Weight 176.22
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Benzyl Methacrylate (stabilized with MEHQ)
EINECSNumber (3)-1017&(3)-1047
TSCA Yes