missing translation for 'onlineSavingsMsg'
Learn More

Benzyl methacrylate, 98%, stab. with ca 50-100ppm 4-methoxyphenol

Catalog No. AAL1084018
Change view
Click to view available options
Quantity:
50 g
250 g
1000 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1084018 50 g
AAL1084030 250 g
AAL108400B 1000 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAL1084018 Supplier Thermo Scientific Chemicals Supplier No. L1084018

CAS: 2495-37-6 | C11H12O2 | 176.215 g/mol

Benzyl methacrylate is an important raw material and intermediate used in organic synthesis, pharmaceuticals and agrochemicals.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Benzyl methacrylate is an important raw material and intermediate used in organic synthesis, pharmaceuticals and agrochemicals.

Solubility
Insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. It is sensitive to light and heat. Incompatible with oxidizing agents.

Chemical Identifiers

CAS 2495-37-6
Molecular Formula C11H12O2
Molecular Weight (g/mol) 176.215
MDL Number MFCD00053684
InChI Key AOJOEFVRHOZDFN-UHFFFAOYSA-N
Synonym benzyl methacrylate, 2-propenoic acid, 2-methyl-, phenylmethyl ester, benzylmethacrylate, methacrylic acid benzyl ester, methacrylic acid, benzyl ester, unii-z3248k2ssm, phenylmethyl 2-methylprop-2-enoate, poly benzylmethacrylate, bzma, methacrylic acid benzyl
PubChem CID 17236
IUPAC Name benzyl 2-methylprop-2-enoate
SMILES CC(=C)C(=O)OCC1=CC=CC=C1

Specifications

Density 1.04
Boiling Point 231°C to 233°C
Flash Point 107°C (225°F)
Refractive Index 1.514
Quantity 50 g
Beilstein 2045924
Sensitivity Light sensitive
Solubility Information Insoluble in water.
Formula Weight 176.22
Percent Purity 98%
Chemical Name or Material Benzyl methacrylate, stabilized with ≈50 to 100ppm 4-methoxyphenol
Show More Show Less
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 219-674-4
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only