missing translation for 'onlineSavingsMsg'
Learn More

Benzyl L-(+)-Mandelate 98.0+%, TCI America™

Catalog No. M13555G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M13555G 5 g
M135525G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M13555G Supplier TCI America Supplier No. M13555G
Only null left

Chemical Identifiers

CAS 62173-99-3
Molecular Formula C15H14O3
Molecular Weight (g/mol) 242.274
MDL Number MFCD00274325
InChI Key JFKWZVQEMSKSBU-AWEZNQCLSA-N
Synonym L-(+)-Mandelic Acid Benzyl Ester
PubChem CID 667423
IUPAC Name benzyl (2S)-2-hydroxy-2-phenylacetate
SMILES C1=CC=C(C=C1)COC(=O)C(C2=CC=CC=C2)O

Specifications

Melting Point 107°C
Color White
Quantity 5 g
Formula Weight 242.27
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Benzyl L-(+)-Mandelate