missing translation for 'onlineSavingsMsg'
Learn More

Benzyl L-lactate, 97%

Catalog No. p-7058424
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1464506 5 g
AAL1464514 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL1464506 Supplier Thermo Scientific Chemicals Supplier No. L1464506

CAS: 56777-24-3 | C10H12O3 | 180.203 g/mol

Benzyl L-lactate is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuffs.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Benzyl L-lactate is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuffs.

Solubility
Not miscible or difficult to mix in water.

Notes
Keep container tightly closed in a dry and well-ventilated place. Containers which are opened must be carefully resealed and kept upright to prevent leakage. Store in cool place. Incompatible with oxidizing agents.

Chemical Identifiers

CAS 56777-24-3
Molecular Formula C10H12O3
Molecular Weight (g/mol) 180.203
MDL Number MFCD00209546
InChI Key ZYTLPUIDJRKAAM-QMMMGPOBSA-N
Synonym benzyl s---lactate, benzyl l-lactate, benzyl s-lactate, s---lactic acid benzyl ester, benzyl 2s-2-hydroxypropanoate, benzyl-l-lactate, s-benzyllactate, s-benzyl lactate, s-lactic acid benzyl ester, 2s-lactic acid benzyl ester
PubChem CID 11052238
IUPAC Name benzyl (2S)-2-hydroxypropanoate
SMILES CC(C(=O)OCC1=CC=CC=C1)O

Specifications

Density 1.12
Boiling Point 105°C to 109°C (0.1 mmHg)
Flash Point >110°C (230°F)
Refractive Index 1.513
Quantity 5 g
Beilstein 2048925
Solubility Information Not miscible or difficult to mix in water.
Optical Rotation −12.9° (c=3.1 in Methanol)
Formula Weight 180.2
Percent Purity 98%
Chemical Name or Material Benzyl L-lactate
Show More Show Less
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only