missing translation for 'onlineSavingsMsg'
Learn More

Benzyl Isocyanide 96.0+%, TCI America™

Catalog No. p-7119535
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B21851G 1 g
B21855G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B21851G Supplier TCI America Supplier No. B21851G
Only null left

Chemical Identifiers

CAS 10340-91-7
Molecular Formula C8H7N
Molecular Weight (g/mol) 117.15
MDL Number MFCD00000004
InChI Key RIWNFZUWWRVGEU-UHFFFAOYSA-N
Synonym benzyl isocyanide, isocyanomethyl benzene, benzylisocyanide, benzene, isocyanomethyl, benzyl isonitrile, benzylimino methylidene, benzylisonitrile, benzylisonit-rile, isocyanomethylbenzol, isocyanomethyl-benzene
PubChem CID 82558
IUPAC Name (isocyanomethyl)benzene
SMILES [C-]#[N+]CC1=CC=CC=C1

Specifications

Boiling Point 106°C
Quantity 1 g
Formula Weight 117.15
Percent Purity ≥96.0% (GC)
Physical Form Liquid
Chemical Name or Material Benzyl Isocyanide
TSCA No
Recommended Storage Freezer