missing translation for 'onlineSavingsMsg'
Learn More

Benzyl isocyanide, 98%

Catalog No. AC158390010
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC158390010 1 g Glass bottle
AC158390050 5 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC158390010 Supplier Thermo Scientific Chemicals Supplier No. 158390010
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 10340-91-7 | C8H7N | 117.15 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 10340-91-7
Molecular Formula C8H7N
Molecular Weight (g/mol) 117.15
MDL Number MFCD00000004
InChI Key RIWNFZUWWRVGEU-UHFFFAOYSA-N
Synonym benzyl isocyanide, isocyanomethyl benzene, benzylisocyanide, benzene, isocyanomethyl, benzyl isonitrile, benzylimino methylidene, benzylisonitrile, benzylisonit-rile, isocyanomethylbenzol, isocyanomethyl-benzene
PubChem CID 82558
IUPAC Name isocyanomethylbenzene
SMILES [C-]#[N+]CC1=CC=CC=C1

Specifications

CAS Min % 97.5
CAS Max % 100.0
Density 0.9620g/mL
Boiling Point 105.0°C to 106.0°C (75.0 mmHg)
Flash Point 79°C
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Refractive Index 1.5200 to 1.5220
Linear Formula C6H5CH2NC
Quantity 1 g
Beilstein 12,1041
Specific Gravity 0.962
Formula Weight 117.18
Percent Purity 98%
Chemical Name or Material Benzyl isocyanide, 98%
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Precautionary Statement GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 233-738-9

RUO – Research Use Only