missing translation for 'onlineSavingsMsg'
Learn More

Benzyl isocyanate, 98%

Catalog No. aal0263306
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0263306 5 g
AAL0263303 1 g
AAL0263314 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAL0263306 Supplier Thermo Scientific Chemicals Supplier No. L0263306

CAS: 3173-56-6 | C8H7NO | 133.15 g/mol

Benzyl isocyanate, is used as a protected ammonia equivalent in the stereo selective ring-opening of chiral 2,3-epoxy alcohols (from Sharpless asymmetric epoxidation). The key step in the sequence involves intramolecular cyclization of the carbamate intermediate. It is also used in a multicomponent, stereospecific synthesis of 1,3-oxazinane-2,4-diones employing an Al-salph complex.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Benzyl isocyanate, is used as a protected ammonia equivalent in the stereo selective ring-opening of chiral 2,3-epoxy alcohols (from Sharpless asymmetric epoxidation). The key step in the sequence involves intramolecular cyclization of the carbamate intermediate. It is also used in a multicomponent, stereospecific synthesis of 1,3-oxazinane-2,4-diones employing an Al-salph complex.

Solubility
It hydrolyzes with water.

Notes
Moisture& heat Sensitive. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Keep away from alcohols, amines.

Chemical Identifiers

CAS 3173-56-6
Molecular Formula C8H7NO
Molecular Weight (g/mol) 133.15
MDL Number MFCD00009701
InChI Key YDNLNVZZTACNJX-UHFFFAOYSA-N
Synonym benzyl isocyanate, isocyanatomethyl benzene, benzylisocyanate, isocyanic acid benzyl ester, benzene, isocyanatomethyl, 1-isocyanatomethyl benzene, phenylmethyl isocyanate, unii-8i2li357gq, phenylmethanisocyanate
PubChem CID 76639
IUPAC Name isocyanatomethylbenzene
SMILES O=C=NCC1=CC=CC=C1

Specifications

Density 1.078
Boiling Point 53°C to 54°C (0.8 mmHg)
Flash Point 43°C (109°F)
Odor Pungent
Refractive Index 1.525
Linear Formula C6H5CH2NCO
Quantity 5 g
UN Number UN3080
Beilstein 636508
Sensitivity Moisture sensitive
Solubility Information It hydrolyzes with water.
Formula Weight 133.15
Percent Purity 98%
Chemical Name or Material Benzyl isocyanate
Show More Show Less
Hazard Category H226-H301+H311+H331-H315-H317-H319-H334-H335
Hazard Statement GHS H Statement
H301-H330-H334-H226-H315-H319-H335
Toxic if swallowed.
Fatal if inhaled.
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Flammable liquid and vapor.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P210-P233-P235-P240-P241-P242-P243-P261-P264b-P270-P271-P272-P280-P285-P301+P310-P303+P361+P353-P304+P340-P305+P351+P338-P311-P312-P330-P333+P313-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: ISOCYANATES, TOXIC, FLAMMABLE, N.O.S.
EINECSNumber 221-640-9
TSCA No
Recommended Storage Keep cold

RUO – Research Use Only