missing translation for 'onlineSavingsMsg'
Learn More

Benzyl glycinate hydrochloride, 98%

Catalog No. AAH5024703
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH5024703 1 g
AAH5024706 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH5024703 Supplier Thermo Scientific Chemicals Supplier No. H5024703
Only null left

CAS: 2462-31-9 | C9H12ClNO2 | 201.65 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 2462-31-9
Molecular Formula C9H12ClNO2
Molecular Weight (g/mol) 201.65
MDL Number MFCD00035442
InChI Key VLQHNAMRWPQWNK-UHFFFAOYSA-N
Synonym benzyl glycinate hydrochloride, benzyl 2-aminoacetate hydrochloride, glycine benzyl ester hydrochloride, h-dl-gly-obzl.hcl, benzyl glycinate hcl, h-gly-obzl.hcl, benzyl glycinate, h-gly-obzl hcl, glycine benzyl ester hcl, glycine, phenylmethyl ester, hydrochloride
PubChem CID 11701227
IUPAC Name benzyl 2-aminoacetate;hydrochloride
SMILES C1=CC=C(C=C1)COC(=O)CN.Cl

Specifications

Melting Point 139°C to 140°C
Linear Formula H2NCH2CO2CH2C6H5·HCl
Quantity 1 g
Formula Weight 201.65
Percent Purity 98%
Chemical Name or Material Benzyl glycinate hydrochloride
TSCA No
Recommended Storage Keep cold

RUO – Research Use Only