missing translation for 'onlineSavingsMsg'
Learn More

Benzyl ethyl malonate, tech. 85%

Catalog No. p-7026790
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2301114 25 g
AAB2301122 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAB2301114 Supplier Thermo Scientific Chemicals Supplier No. B2301114

CAS: 42998-51-6 | C12H14O4 | 222.24 g/mol

Benzyl ethyl malonate is used as a pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Benzyl ethyl malonate is used as a pharmaceutical intermediate.

Solubility
Soluble in water 525.7 mg/L at 25°C. Soluble in alcohol.

Notes
Store in cool, dry place in a well sealed container. Store away from oxidizing agents.

Chemical Identifiers

CAS 42998-51-6
Molecular Formula C12H14O4
Molecular Weight (g/mol) 222.24
MDL Number MFCD00009194
InChI Key CGNOCUSLPSCMLL-UHFFFAOYSA-N
Synonym benzyl ethyl malonate, benzylethyl malonate, malonic acid benzyl ethyl ester, ethyl benzylmalonate, acmc-20alpf, 1-benzyl 3-ethyl malonate #, ethyl phenylmethyl propanedioate, 1-benzyl 3-ethyl propanedioate, malonic acid 1-benzyl 3-ethyl ester, ethyl phenylmethyl propane-1,3-dioate
PubChem CID 562228
SMILES CCOC(=O)CC(=O)OCC1=CC=CC=C1

Specifications

Boiling Point 145°C (4 mmHg)
Refractive Index 1.497
Quantity 25 g
Solubility Information Soluble in water 525.7mg/L @ 25°C. Soluble in alcohol.
IUPAC Name 1-benzyl 3-ethyl propanedioate
Formula Weight 222.24
Percent Purity 85%
Grade Technical
Chemical Name or Material Benzyl ethyl malonate
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only