missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Benzyl D-(-)-Mandelate 98.0+%, TCI America™
$63.38 - $188.27
Chemical Identifiers
| CAS | 97415-09-3 |
|---|---|
| Molecular Formula | C15H14O3 |
| Molecular Weight (g/mol) | 242.274 |
| MDL Number | MFCD00674031 |
| InChI Key | JFKWZVQEMSKSBU-CQSZACIVSA-N |
| Synonym | D-(-)-Mandelic Acid Benzyl Ester |
| PubChem CID | 853755 |
| IUPAC Name | benzyl (2R)-2-hydroxy-2-phenylacetate |
| SMILES | C1=CC=C(C=C1)COC(=O)C(C2=CC=CC=C2)O |
Chemical Identifiers
| 97415-09-3 | |
| 242.274 | |
| JFKWZVQEMSKSBU-CQSZACIVSA-N | |
| 853755 | |
| C1=CC=C(C=C1)COC(=O)C(C2=CC=CC=C2)O |
| C15H14O3 | |
| MFCD00674031 | |
| D-(-)-Mandelic Acid Benzyl Ester | |
| benzyl (2R)-2-hydroxy-2-phenylacetate |
Specifications
| 97415-09-3 | |
| White-Yellow | |
| MFCD00674031 | |
| D-(-)-Mandelic Acid Benzyl Ester | |
| C1=CC=C(C=C1)COC(=O)C(C2=CC=CC=C2)O | |
| 242.274 | |
| 242.27 | |
| Crystalline Powder |
| 107°C | |
| C15H14O3 | |
| 5 g | |
| JFKWZVQEMSKSBU-CQSZACIVSA-N | |
| benzyl (2R)-2-hydroxy-2-phenylacetate | |
| 853755 | |
| ≥98.0% (GC) | |
| Benzyl D-(-)-Mandelate |
Safety and Handling
TSCA : No