missing translation for 'onlineSavingsMsg'
Learn More

Benzyl cyanoacetate, 97%

CAS: 14447-18-8 | C10H9NO2 | 175.19 g/mol

$148.12 - $466.04

Chemical Identifiers

CAS 14447-18-8
Molecular Formula C10H9NO2
Molecular Weight (g/mol) 175.19
MDL Number MFCD00051719
InChI Key RCUIWQWWDLZNMS-UHFFFAOYSA-N
Synonym benzyl cyanoacetate, acetic acid, cyano-, phenylmethyl ester, cyanoacetic acid benzyl ester, benzyl-cyanoacetate, acetic acid, cyano-, benzyl ester, acmc-20alph, phenylmethyl 2-cyanoacetate, cyano-acetic acid benzyl ester, benzyl ester of cyanoacetic acid
PubChem CID 560818
IUPAC Name benzyl 2-cyanoacetate
SMILES O=C(CC#N)OCC1=CC=CC=C1
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAL0664414
View Documents
Thermo Scientific Chemicals
L0664414
25 g
Each for $148.12
Only null left
 
AAL0664422
View Documents
Thermo Scientific Chemicals
L0664422
100 g
Each for $466.04
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

14447-18-8
175.19
RCUIWQWWDLZNMS-UHFFFAOYSA-N
560818
O=C(CC#N)OCC1=CC=CC=C1
C10H9NO2
MFCD00051719
benzyl cyanoacetate, acetic acid, cyano-, phenylmethyl ester, cyanoacetic acid benzyl ester, benzyl-cyanoacetate, acetic acid, cyano-, benzyl ester, acmc-20alph, phenylmethyl 2-cyanoacetate, cyano-acetic acid benzyl ester, benzyl ester of cyanoacetic acid
benzyl 2-cyanoacetate

Specifications

14447-18-8
140°C (0.8 mmHg)
Odorless
1.5195
25 g
benzyl cyanoacetate, acetic acid, cyano-, phenylmethyl ester, cyanoacetic acid benzyl ester, benzyl-cyanoacetate, acetic acid, cyano-, benzyl ester, acmc-20alph, phenylmethyl 2-cyanoacetate, cyano-acetic acid benzyl ester, benzyl ester of cyanoacetic acid
O=C(CC#N)OCC1=CC=CC=C1
175.19
175.18
Benzyl cyanoacetate
26°C to 30°C
>100°C (212°F)
C10H9NO2
MFCD00051719
2096578
RCUIWQWWDLZNMS-UHFFFAOYSA-N
benzyl 2-cyanoacetate
560818
97%
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H301
Toxic if swallowed.

P264b-P270-P301+P312-P330-P501c

H302

TSCA : No

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only