missing translation for 'onlineSavingsMsg'
Learn More

Benzyl cyanoacetate, 97%

Catalog No. AAL0664414
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0664414 25 g
AAL0664422 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL0664414 Supplier Thermo Scientific Chemicals Supplier No. L0664414
Only null left

CAS: 14447-18-8 | C10H9NO2 | 175.19 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 14447-18-8
Molecular Formula C10H9NO2
Molecular Weight (g/mol) 175.19
MDL Number MFCD00051719
InChI Key RCUIWQWWDLZNMS-UHFFFAOYSA-N
Synonym benzyl cyanoacetate, acetic acid, cyano-, phenylmethyl ester, cyanoacetic acid benzyl ester, benzyl-cyanoacetate, acetic acid, cyano-, benzyl ester, acmc-20alph, phenylmethyl 2-cyanoacetate, cyano-acetic acid benzyl ester, benzyl ester of cyanoacetic acid
PubChem CID 560818
IUPAC Name benzyl 2-cyanoacetate
SMILES O=C(CC#N)OCC1=CC=CC=C1

Specifications

Melting Point 26°C to 30°C
Boiling Point 140°C (0.8 mmHg)
Flash Point >100°C (212°F)
Odor Odorless
Refractive Index 1.5195
Quantity 25 g
Beilstein 2096578
Formula Weight 175.18
Percent Purity 97%
Chemical Name or Material Benzyl cyanoacetate
Hazard Category H302
Hazard Statement GHS H Statement
H301
Toxic if swallowed.
Precautionary Statement P264b-P270-P301+P312-P330-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only