missing translation for 'onlineSavingsMsg'
Learn More

Benzyl Cinnamate 98.0+%, TCI America™

Catalog No. C035825G
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C035825G 25 g
C0358500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C035825G Supplier TCI America Supplier No. C035825G

Chemical Identifiers

CAS 103-41-3
Molecular Formula C16H14O2
Molecular Weight (g/mol) 238.29
MDL Number MFCD00004789,MFCD00004789,MFCD00004789
InChI Key NGHOLYJTSCBCGC-VAWYXSNFSA-N
Synonym benzyl alcohol cinnamic ester, benzylcinnamate, benzyl alcohol, cinnamate, wln: r1u1vo1r, benzyl .gamma.-phenylacrylate, benzyl cis-cinnamate, benzyl z-cinnamate, 2-propenoic acid, 3-phenyl-, phenylmethyl ester, z-benzyl 3-phenylacrylate, cinnamic acid, benzyl ester, z
PubChem CID 15558051
IUPAC Name benzyl (2E)-3-phenylprop-2-enoate
SMILES O=C(OCC1=CC=CC=C1)\C=C\C1=CC=CC=C1

Specifications

Melting Point 36°C
Color White-Yellow
Boiling Point 230°C
Quantity 25 g
Formula Weight 238.29
Percent Purity ≥98.0% (GC)
Physical Form Crystals
Chemical Name or Material Benzyl Cinnamate
EINECSNumber (3)-1739
RTECSNumber GD8400000
TSCA Yes