missing translation for 'onlineSavingsMsg'
Learn More

Benzyl Chloroformate 96.0+%, TCI America™

Supplier:  TCI America B302125G

 View more versions of this product

Catalog No. B302125G


Only null left
Explore available promotions

Description

Description

Specifications

Specifications

Benzyl Chloroformate
0°C
103°C
MFCD00000640
1739
HSDAJNMJOMSNEV-UHFFFAOYSA-N
benzyl carbonochloridate
10387
≥96.0% (T)
501-53-1
Yellow
C8H7ClO2
25 g
benzyl chloroformate, carbobenzoxy chloride, benzyl chlorocarbonate, cbz chloride, benzyloxycarbonyl chloride, carbobenzyloxy chloride, carbonochloridic acid, phenylmethyl ester, benzylcarbonyl chloride, cbz-cl, chloroformic acid, benzyl ester
C1=CC=C(C=C1)COC(=O)Cl
170.592
170.59
Liquid

Chemical Identifiers

501-53-1
170.592
HSDAJNMJOMSNEV-UHFFFAOYSA-N
10387
C1=CC=C(C=C1)COC(=O)Cl
C8H7ClO2
MFCD00000640
benzyl chloroformate, carbobenzoxy chloride, benzyl chlorocarbonate, cbz chloride, benzyloxycarbonyl chloride, carbobenzyloxy chloride, carbonochloridic acid, phenylmethyl ester, benzylcarbonyl chloride, cbz-cl, chloroformic acid, benzyl ester
benzyl carbonochloridate
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

EINECSNumber : (3)-1835&(3)-2885

RTECSNumber : LQ5860000

TSCA : Yes

Recommended Storage : Refrigerator

SDS
Documents

Documents

Promotions

Promotions