missing translation for 'onlineSavingsMsg'
Learn More

Benzyl Chloroformate 96.0+%, TCI America™

Catalog No. B3021250G
Change view
Click to view available options
Quantity:
25 g
250 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B3021250G 250 g
B302125G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B3021250G Supplier TCI America Supplier No. B3021250G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 501-53-1
Molecular Formula C8H7ClO2
Molecular Weight (g/mol) 170.592
MDL Number MFCD00000640
InChI Key HSDAJNMJOMSNEV-UHFFFAOYSA-N
Synonym benzyl chloroformate, carbobenzoxy chloride, benzyl chlorocarbonate, cbz chloride, benzyloxycarbonyl chloride, carbobenzyloxy chloride, carbonochloridic acid, phenylmethyl ester, benzylcarbonyl chloride, cbz-cl, chloroformic acid, benzyl ester
PubChem CID 10387
IUPAC Name benzyl carbonochloridate
SMILES C1=CC=C(C=C1)COC(=O)Cl

Specifications

Melting Point 0°C
Color Yellow
Boiling Point 103°C
Quantity 250 g
UN Number 1739
Formula Weight 170.59
Percent Purity ≥96.0% (T)
Physical Form Liquid
Chemical Name or Material Benzyl Chloroformate
EINECSNumber (3)-1835&(3)-2885
RTECSNumber LQ5860000
TSCA Yes
Recommended Storage Refrigerator