missing translation for 'onlineSavingsMsg'
Learn More

Benzyl chloroformate, 97 wt%, stabilized

Catalog No. p-4510242
Change view
Click to view available options
Quantity:
100 g
500 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC152941000 100 g Glass bottle
AC152945000 500 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC152941000 Supplier Thermo Scientific Chemicals Supplier No. 152941000

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Benzyl chloroformate, 97 wt%, stabilized, Quantity: 500g, Packaging: Glass bottle, Boiling Point: 178.0 deg.C to 180.0 deg.C, Undesignated, Melting Point: -20.0 deg.C, Molecular Weight: 170.6, Percent Purity: 97 wt%, Assay Percent Range: 96% min. (GC), Beilstein: 06,437 | CAS: 501-53-1 | C8H7ClO2 | 170.6 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Coupling reagent

Chemical Identifiers

CAS 501-53-1 , 100-44-7
Molecular Formula C8H7ClO2
Molecular Weight (g/mol) 170.6
InChI Key HSDAJNMJOMSNEV-UHFFFAOYSA-N
Synonym benzyl chloroformate, carbobenzoxy chloride, benzyl chlorocarbonate, cbz chloride, benzyloxycarbonyl chloride, carbobenzyloxy chloride, carbonochloridic acid, phenylmethyl ester, benzylcarbonyl chloride, cbz-cl, chloroformic acid, benzyl ester
PubChem CID 10387
IUPAC Name benzyl carbonochloridate
SMILES C1=CC=C(C=C1)COC(=O)Cl

Specifications

Melting Point -20.0°C
Density 1.2000g/mL
Boiling Point 178.0°C to 180.0°C
Flash Point 92°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Refractive Index 1.5160 to 1.5180
Linear Formula ClCO2CH2C6H5
Quantity 100 g
Beilstein 06,437
Fieser 01,109; 02,59; 15,22
Merck Index 15,1801
Specific Gravity 1.2
Solubility Information Solubility in water: reacts
Stabilizer 0.1% anhydrous sodium carbonate
Viscosity 2.36 mPa.s (20°C)
Formula Weight 170.6
Percent Purity 97 wt%
Physical Form Liquid
Chemical Name or Material Benzyl chloroformate
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
May be corrosive to metals.
Causes severe skin burns and eye damage.
Harmful if inhaled.
May cause cancer.
Very toxic to aquatic life with long lasting effects.
Precautionary Statement GHS P Statement
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if pre
EINECSNumber 207-925-
Recommended Storage Important Note: the material builds progressively benzyl chloride under normal storage conditions

RUO – Research Use Only