missing translation for 'onlineSavingsMsg'
Learn More

Benzyl chloroformate, 3M (50 wt%) solution in toluene

CAS: 501-53-1 | C8H7ClO2 | 170.6 g/mol

$388.63 - $388.63

Specifications

Chemical Name or Material Benzyl chloroformate
Name Note 50 wt. % Solution in Toluene
CAS 501-53-1,108-88-3,100-44-7
CAS Min % 45.0,47.0
CAS Max % 53.0,55.0
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC124495000
View Documents
Thermo Scientific Chemicals
124495000
500 mL Glass bottle
Each for $388.63
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Coupling reagent
Specifications

Specifications

Benzyl chloroformate
501-53-1,108-88-3,100-44-7
53.0,55.0
7°C
C8H7ClO2
MFCD00000640
01,109; 02,59; 15,22
1.01
Solubility in water: decomposes
C1=CC=C(C=C1)COC(=O)Cl
170.6
170.6
Solution
50 wt. % Solution in Toluene
45.0,47.0
1.0100g/mL
Glass bottle
C6H5CH2OOCCl
06, 437
15, 1801
benzyl chloroformate, carbobenzoxy chloride, benzyl chlorocarbonate, cbz chloride, benzyloxycarbonyl chloride, carbobenzyloxy chloride, carbonochloridic acid, phenylmethyl ester, benzylcarbonyl chloride, cbz-cl, chloroformic acid, benzyl ester
HSDAJNMJOMSNEV-UHFFFAOYSA-N
benzyl carbonochloridate
10387
45 to 53 wt%
Videos
SDS
Documents

Documents

RUO – Research Use Only