missing translation for 'onlineSavingsMsg'
Learn More

Benzyl chloroformate, 3M (50 wt.%) solution in toluene, AcroSeal™

Catalog No. AC428901000
Change view
Click to view available options
Quantity:
100 mL
Packaging:
AcroSeal™ Glass bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC428901000 100 mL AcroSeal™ Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC428901000 Supplier Thermo Scientific Chemicals Supplier No. 428901000

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 501-53-1 | C8H7ClO2 | 170.6 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Coupling reagent

Specifications

Chemical Name or Material Benzyl chloroformate
CAS 501-53-1, 108-88-3, 100-44-7
CAS Min % 49.83, 44.26
CAS Max % 55.74, 50.17
Color Yellow to Pink
Density 1.0100g/mL
Flash Point 7°C
Packaging AcroSeal™ Glass bottle
Molecular Formula C8H7ClO2
Linear Formula C6H5CH2OOCCl
Quantity 100 mL
MDL Number MFCD00000640
Specific Gravity 1.01
Merck Index 15, 1801
Synonym benzyl chloroformate, carbobenzoxy chloride, benzyl chlorocarbonate, cbz chloride, benzyloxycarbonyl chloride, carbobenzyloxy chloride, carbonochloridic acid, phenylmethyl ester, benzylcarbonyl chloride, cbz-cl, chloroformic acid, benzyl ester
Solubility Information Solubility in water: decomposes
InChI Key HSDAJNMJOMSNEV-UHFFFAOYSA-N
SMILES C1=CC=C(C=C1)COC(=O)Cl
IUPAC Name benzyl carbonochloridate
Molecular Weight (g/mol) 170.6
PubChem CID 10387
Formula Weight 170.6
Percent Purity 45 to 53 wt%
Physical Form Solution
Show More Show Less

RUO – Research Use Only