missing translation for 'onlineSavingsMsg'
Learn More

Benzyl Chloroformate (30-35% in Toluene), TCI America™

Catalog No. C0176500ML
Change view
Click to view available options
Quantity:
25 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C0176500ML 500 mL
C017625ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C0176500ML Supplier TCI America Supplier No. C0176500ML

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 501-53-1
Molecular Formula C8H7ClO2
Molecular Weight (g/mol) 170.592
MDL Number MFCD00000640
InChI Key HSDAJNMJOMSNEV-UHFFFAOYSA-N
Synonym benzyl chloroformate, carbobenzoxy chloride, benzyl chlorocarbonate, cbz chloride, benzyloxycarbonyl chloride, carbobenzyloxy chloride, carbonochloridic acid, phenylmethyl ester, benzylcarbonyl chloride, cbz-cl, chloroformic acid, benzyl ester
PubChem CID 10387
IUPAC Name benzyl carbonochloridate
SMILES C1=CC=C(C=C1)COC(=O)Cl

Specifications

Color Colorless
Quantity 500 mL
UN Number 2920
Formula Weight 170.59
Physical Form Liquid
Chemical Name or Material Benzyl Chloroformate (30-35% in Toluene)
EINECSNumber (3)-1835
RTECSNumber LQ5860000
TSCA Yes