missing translation for 'onlineSavingsMsg'
Learn More

Benzyl Butyrate 97.0+%, TCI America™

Catalog No. b0756500g
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B0756500G 500 g
B075625G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B0756500G Supplier TCI America Supplier No. B0756500G
Only null left

Chemical Identifiers

CAS 103-37-7
Molecular Formula C11H14O2
Molecular Weight (g/mol) 178.231
MDL Number MFCD00027133
InChI Key VONGZNXBKCOUHB-UHFFFAOYSA-N
Synonym benzyl butyrate, benzyl n-butyrate, butanoic acid, phenylmethyl ester, phenylmethyl butanoate, benzyl n-butanoate, butyric acid, benzyl ester, phenylmethyl butyrate, benzylester kyseliny maselne, benzylbutyrate, benzyl butyrate natural
PubChem CID 7650
IUPAC Name benzyl butanoate
SMILES CCCC(=O)OCC1=CC=CC=C1

Specifications

Color Yellow
Boiling Point 240°C
Quantity 500 g
Formula Weight 178.23
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material Benzyl Butyrate
EINECSNumber (3)-1020&(3)-1045
RTECSNumber ES7350000
TSCA Yes