missing translation for 'onlineSavingsMsg'
Learn More

Benzyl Bromoacetate 96.0+%, TCI America™

Catalog No. B167425G
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B167425G 25 g
B1674500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B167425G Supplier TCI America Supplier No. B167425G

Chemical Identifiers

CAS 5437-45-6
Molecular Formula C9H9BrO2
Molecular Weight (g/mol) 229.073
MDL Number MFCD00000190
InChI Key JHVLLYQQQYIWKX-UHFFFAOYSA-N
Synonym benzyl bromoacetate, bromoacetic acid benzyl ester, acetic acid, bromo-, phenylmethyl ester, merbac 35, caswell no. 082a, acetic acid, bromo-, benzyl ester, unii-64u2rk18d3, phenylmethyl 2-bromoacetate, epa pesticide chemical code 008710, benzyl bromacetate
PubChem CID 62576
IUPAC Name benzyl 2-bromoacetate
SMILES C1=CC=C(C=C1)COC(=O)CBr

Specifications

Color Yellow
Boiling Point 170°C
Quantity 25 g
Formula Weight 229.07
Percent Purity ≥96.0% (GC)
Physical Form Liquid
Chemical Name or Material Benzyl Bromoacetate
EINECSNumber (9)-1875
RTECSNumber AF5957215
TSCA No