missing translation for 'onlineSavingsMsg'
Learn More

Benzyl bromoacetate, 97%

Catalog No. AAA1763030
Change view
Click to view available options
Quantity:
50 g
250 g
1000 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1763030 250 g
AAA1763018 50 g
AAA176300B 1000 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1763030 Supplier Thermo Scientific Chemicals Supplier No. A1763030
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 5437-45-6 | C9H9BrO2 | 229.073 g/mol

It is employed in biological research purpose. Benzyl bromoacetate was used in the alkylation of (-)-2,3-O-isopropylidene-D-threitol that afforded lipopeptide, 2-[(4R,5R)-5-({[(9H-fluoren-9-yl)methoxy]carbonylaminomethyl}-2,2-dimethyl-1,3-dioxolan-4-yl)methoxy]acetic acid.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is employed in biological research purpose. Benzyl bromoacetate was used in the alkylation of (-)-2,3-O-isopropylidene-D-threitol that afforded lipopeptide, 2-[(4R,5R)-5-({[(9H-fluoren-9-yl)methoxy]carbonylaminomethyl}-2,2-dimethyl-1,3-dioxolan-4-yl)methoxy]acetic acid.

Solubility
Not miscible or difficult to mix in water.

Notes
Store away from oxidizing agents and strong bases. Keep the container tightly closed in a cool, dry place. Store under 20˚C Freezer, Under Inert Atmosphere.

Chemical Identifiers

CAS 5437-45-6
Molecular Formula C9H9BrO2
Molecular Weight (g/mol) 229.073
MDL Number MFCD00000190
InChI Key JHVLLYQQQYIWKX-UHFFFAOYSA-N
Synonym benzyl bromoacetate, bromoacetic acid benzyl ester, acetic acid, bromo-, phenylmethyl ester, merbac 35, caswell no. 082a, acetic acid, bromo-, benzyl ester, unii-64u2rk18d3, phenylmethyl 2-bromoacetate, epa pesticide chemical code 008710, benzyl bromacetate
PubChem CID 62576
IUPAC Name benzyl 2-bromoacetate
SMILES C1=CC=C(C=C1)COC(=O)CBr

Specifications

Density 1.46
Boiling Point 157°C to 159°C (10 mmHg)
Flash Point >110°C (230°F)
Odor Pungent
Refractive Index 1.544
Quantity 250 g
Beilstein 973658
Solubility Information Not miscible or difficult to mix in water.
Formula Weight 229.08
Percent Purity 97%
Chemical Name or Material Benzyl bromoacetate
Show More Show Less
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 226-611-4
RTECSNumber AF5957215
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only