missing translation for 'onlineSavingsMsg'
Learn More

Benzyl acrylate, 98%, stab. with ca 150ppm 4-methoxyphenol

Catalog No. AA4320409
Change view
Click to view available options
Quantity:
10 g
50 g
250 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA4320409 10 g
AA4320418 50 g
AA4320430 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AA4320409 Supplier Thermo Scientific Chemicals Supplier No. 04320409

Preparation of high refractive index polyacrylates | CAS: 2495-35-4 | C10H10O2 | 162.19 g/mol

Preparation of high refractive index polyacrylates.Benzyl acrylate is used in the preparation of heptanoic acid benzyl ester. It is used to prepare polybenzylacrylate using azobisisobutyronitrile as initiator.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 2495-35-4
Molecular Formula C10H10O2
Molecular Weight (g/mol) 162.19
MDL Number MFCD00048147
InChI Key GCTPMLUUWLLESL-UHFFFAOYSA-N
Synonym benzyl acrylate, benzylacrylate, 2-propenoic acid, phenylmethyl ester, sartomer sr 432, acrylic acid, benzyl ester, acrylic acid benzyl ester, phenylmethyl prop-2-enoate, benzyl 2-propenoate, fancryl fa-bza, acrylic acid benzyl
PubChem CID 75617
IUPAC Name benzyl prop-2-enoate
SMILES C=CC(=O)OCC1=CC=CC=C1

Specifications

Color Colorless
Density 1.06 g/mL
Boiling Point 111°C (8 mmHg)
Refractive Index 1.518
Linear Formula C6H5CH2CO2CH=CH2
Quantity 10 g
UN Number UN3082
Solubility Information Miscible with organic solvents and water.
Formula Weight 162.19
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material Benzyl acrylate, Stabilized with ≈150ppm 4-methoxyphenol
Show More Show Less
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
DOTInformation Transport Hazard Class: 9; Packing Group: III; Proper Shipping Name: ENVIRONMENTALLY HAZARDOUS SUBSTANCE, LIQUID, N.O.S.
EINECSNumber 219-673-9
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only