missing translation for 'onlineSavingsMsg'
Learn More

Benzyl Acetoacetate 95.0+%, TCI America™

Catalog No. A1080500ML
Change view
Click to view available options
Quantity:
25 mL
100 mL
500 mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A1080500ML 500 mL
A108025ML 25 mL
A1080100ML 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. A1080500ML Supplier TCI America Supplier No. A1080500ML

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 5396-89-4
Molecular Formula C11H12O3
Molecular Weight (g/mol) 192.214
MDL Number MFCD00008785
InChI Key WOFAGNLBCJWEOE-UHFFFAOYSA-N
Synonym benzyl acetoacetate, acetoacetic acid benzyl ester, benzyl acetylacetate, acetoacetic acid, benzyl ester, phenylmethyl 3-oxobutanoate, butanoic acid, 3-oxo-, phenylmethyl ester, benzyl beta-ketobutyrate, ac-benzyl, unii-f9s0xgv18x, f9s0xgv18x
PubChem CID 142266
IUPAC Name benzyl 3-oxobutanoate
SMILES CC(=O)CC(=O)OCC1=CC=CC=C1

Specifications

Boiling Point 163°C
Quantity 500 mL
Formula Weight 192.21
Percent Purity ≥95.0% (GC,T)
Physical Form Liquid
Chemical Name or Material Benzyl Acetoacetate