missing translation for 'onlineSavingsMsg'
Learn More

Benzyl Acetoacetate 95.0+%, TCI America™

Catalog No. A108025ML
Change view
Click to view available options
Quantity:
25 mL
100 mL
500 mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A108025ML 25 mL
A1080100ML 100 mL
A1080500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. A108025ML Supplier TCI America Supplier No. A108025ML

Chemical Identifiers

CAS 5396-89-4
Molecular Formula C11H12O3
Molecular Weight (g/mol) 192.214
MDL Number MFCD00008785
InChI Key WOFAGNLBCJWEOE-UHFFFAOYSA-N
Synonym benzyl acetoacetate, acetoacetic acid benzyl ester, benzyl acetylacetate, acetoacetic acid, benzyl ester, phenylmethyl 3-oxobutanoate, butanoic acid, 3-oxo-, phenylmethyl ester, benzyl beta-ketobutyrate, ac-benzyl, unii-f9s0xgv18x, f9s0xgv18x
PubChem CID 142266
IUPAC Name benzyl 3-oxobutanoate
SMILES CC(=O)CC(=O)OCC1=CC=CC=C1

Specifications

Boiling Point 163°C
Quantity 25 mL
Formula Weight 192.21
Percent Purity ≥95.0% (GC,T)
Physical Form Liquid
Chemical Name or Material Benzyl Acetoacetate