missing translation for 'onlineSavingsMsg'
Learn More

Benzyl acetoacetate, 97%

Catalog No. AAA1491514
Change view
Click to view available options
Quantity:
25 g
50 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1491514 25 g
AAA1491518 50 g
AAA1491522 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1491514 Supplier Thermo Scientific Chemicals Supplier No. A1491514
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 5396-89-4 | C11H12O3 | 192.214 g/mol

Benzyl acetoacetate is used as a Pharmaceutical and fine chemical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Benzyl acetoacetate is used as a Pharmaceutical and fine chemical intermediate.

Solubility
It is soluble in alkali solutions.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place.

Chemical Identifiers

CAS 5396-89-4
Molecular Formula C11H12O3
Molecular Weight (g/mol) 192.214
MDL Number MFCD00008785
InChI Key WOFAGNLBCJWEOE-UHFFFAOYSA-N
Synonym benzyl acetoacetate, acetoacetic acid benzyl ester, benzyl acetylacetate, acetoacetic acid, benzyl ester, phenylmethyl 3-oxobutanoate, butanoic acid, 3-oxo-, phenylmethyl ester, benzyl beta-ketobutyrate, ac-benzyl, unii-f9s0xgv18x, f9s0xgv18x
PubChem CID 142266
IUPAC Name benzyl 3-oxobutanoate
SMILES CC(=O)CC(=O)OCC1=CC=CC=C1

Specifications

Density 1.113
Boiling Point 276°C
Flash Point 108°C (226°F)
Refractive Index 1.512
Quantity 25 g
Beilstein 2049615
Solubility Information It is soluble in alkali solutions.
Formula Weight 192.22
Percent Purity 97%
Chemical Name or Material Benzyl acetoacetate
Hazard Category H319
Hazard Statement GHS H Statement
H315-H319
Causes skin irritation.
Causes serious eye irritation.
Precautionary Statement P264b-P280i-P305+P351+P338
EINECSNumber 226-416-4
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only