missing translation for 'onlineSavingsMsg'
Learn More

Benzyl 5-amino-1H-pyrazole-4-carboxylate, 98+%

Catalog No. p-7047176
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1785903 1 g
AAL1785906 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL1785903 Supplier Thermo Scientific Chemicals Supplier No. L1785903

CAS: 32016-28-7 | C11H11N3O2 | 217.23 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 32016-28-7
Molecular Formula C11H11N3O2
Molecular Weight (g/mol) 217.23
MDL Number MFCD03426023
InChI Key QBEUXDWEKIOSIL-UHFFFAOYSA-N
Synonym benzyl 5-aminopyrazole-4-carboxylate, benzyl 3-amino-2h-pyrazole-4-carboxylate, acmc-20alpl, phenylmethyl 5-aminopyrazole-4-carboxylate, 3-amino-1h-pyrazole-4-carboxylic acid benzyl ester, 1h-pyrazole-4-carboxylicacid, 3-amino-, phenylmethyl ester
PubChem CID 4360500
IUPAC Name benzyl 5-amino-1H-pyrazole-4-carboxylate
SMILES NC1=C(C=NN1)C(=O)OCC1=CC=CC=C1

Specifications

Melting Point 121°C to 123°C
Quantity 1 g
Beilstein 791498
Formula Weight 217.23
Percent Purity ≥98%
Chemical Name or Material Benzyl 5-amino-1H-pyrazole-4-carboxylate
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only