missing translation for 'onlineSavingsMsg'
Learn More

Benzyl 4-Nitrophenyl Carbonate 96.0+%, TCI America™

Catalog No. C15915G
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C15915G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. C15915G Supplier TCI America Supplier No. C15915G
Only null left

Chemical Identifiers

CAS 13795-24-9
Molecular Formula C14H11NO5
Molecular Weight (g/mol) 273.24
MDL Number MFCD00007323
InChI Key QIXRWIVDBZJDGD-UHFFFAOYSA-N
Synonym Carbonic Acid Benzyl 4-Nitrophenyl Ester
PubChem CID 83737
IUPAC Name benzyl 4-nitrophenyl carbonate
SMILES [O-][N+](=O)C1=CC=C(OC(=O)OCC2=CC=CC=C2)C=C1

Specifications

Melting Point 76°C
Color White-Yellow
Quantity 5 g
Formula Weight 273.24
Percent Purity ≥96.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Benzyl 4-Nitrophenyl Carbonate