missing translation for 'onlineSavingsMsg'
Learn More

Benzyl 4-(bromomethyl)tetrahydro-1(2H)-pyridinecarboxylate, 97%, Thermo Scientific™

Catalog No. p-2703710
Change view
Click to view available options
Quantity:
250mg
1g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
CC07508CB 250mg
CC07508DA 1g
CC07508DE 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. CC07508CB Supplier Thermo Scientific Chemicals Supplier No. CC07508CB

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 159275-17-9
Molecular Formula C14H18BrNO2
Molecular Weight (g/mol) 312.207
MDL Number MFCD02681989
InChI Key XJHKDSZGAWXUTB-UHFFFAOYSA-N
Synonym benzyl 4-bromomethyl piperidine-1-carboxylate, benzyl 4-bromomethyl tetrahydro-1 2h-pyridinecarboxylate, 1-cbz-4-bromomethyl piperidine, n-cbz-4-bromomethyl piperidine, phenylmethyl 4-bromomethyl piperidinecarboxylate, 1-piperidinecarboxylic acid, 4-bromomethyl-, phenylmethyl ester, benzyloxycarbonylbromomethylpiperidine, n-benzyloxycarbonyl-4-bromomethyl piperidine, benzyl 4-bromoethyl tetrahydro-1 2h-pyridinecarboxylate, benzyl 4-bromomethyl tetrahydro-1 2h pyridinecarboxylate
PubChem CID 2776274
IUPAC Name benzyl 4-(bromomethyl)piperidine-1-carboxylate
SMILES C1CN(CCC1CBr)C(=O)OCC2=CC=CC=C2

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber glass bottle
Quantity 250mg
Formula Weight 312.21
Percent Purity 97%
Chemical Name or Material Benzyl 4-(bromomethyl)tetrahydro-1(2H)-pyridinecarboxylate
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement:
Causes serious eye damage.
Causes severe skin burns and eye damage.
Precautionary Statement GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.