missing translation for 'onlineSavingsMsg'
Learn More

Benzyl 2-Naphthyl Ether 98.0+%, TCI America™

Catalog No. B1619500G
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B1619500G 500 g
B161925G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B1619500G Supplier TCI America Supplier No. B1619500G

Chemical Identifiers

CAS 613-62-7
Molecular Formula C17H14O
Molecular Weight (g/mol) 234.298
MDL Number MFCD00089092
InChI Key WLTCCDHHWYAMCG-UHFFFAOYSA-N
Synonym benzyl 2-naphthyl ether, 2-benzyloxy naphthalene, 2-benzyloxynaphthalene, naphthalene, 2-phenylmethoxy, 2-phenylmethoxy naphthalene, 2-phenylmethoxy-naphthalene, 2-benzyloxy-naphthalene, 2-phenylmethoxy naphthalenen, benzyl2-naphthylether, b-naphthylbenzyl ether
PubChem CID 123080
IUPAC Name 2-phenylmethoxynaphthalene
SMILES C1=CC=C(C=C1)COC2=CC3=CC=CC=C3C=C2

Specifications

Melting Point 102°C
Color White
Quantity 500 g
Formula Weight 234.30
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Benzyl 2-Naphthyl Ether
EINECSNumber (4)-1458
TSCA No