missing translation for 'onlineSavingsMsg'
Learn More

Benzyl 2-Hydroxyphenyl Ketone 98.0+%, TCI America™

Numéro de catalogue. B4673200MG
Change view
Click to view available options
Quantity:
200 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Quantity
B4673200MG 200 mg
B46731G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Numéro de catalogue. B4673200MG Fournisseur TCI America Code fournisseur B4673200MG
Il en reste null

Identifiants chimiques

CAS 2491-31-8
Molecular Formula C14H12O2
Molecular Weight (g/mol) 212.25
MDL Number MFCD00218589
InChI Key VGHVJQXWDXRTRJ-UHFFFAOYSA-N
Synonym 2′C-Hydroxy-2-phenylacetophenone
PubChem CID 137612
IUPAC Name 1-(2-hydroxyphenyl)-2-phenylethan-1-one
SMILES OC1=CC=CC=C1C(=O)CC1=CC=CC=C1

Spécifications

Melting Point 57°C
Boiling Point 165°C
Quantity 200 mg
Formula Weight 212.25
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Benzyl 2-Hydroxyphenyl Ketone