missing translation for 'onlineSavingsMsg'
Learn More

Benzyl 2-(hydroxymethyl)-1-indolinecarboxylate, ≥95%, Thermo Scientific™

Catalog No. p-2703695
Change view
Click to view available options
Quantity:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
CC13409DA 1g
CC13409DE 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. CC13409DA Supplier Thermo Scientific Chemicals Supplier No. CC13409DA

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 135829-04-8
Molecular Formula C17H17NO3
Molecular Weight (g/mol) 283.327
MDL Number MFCD02682013
InChI Key QSMOQTIGILELKY-UHFFFAOYSA-N
Synonym benzyl 2-hydroxymethyl-1-indolinecarboxylate, benzyl 2-hydroxymethyl indoline-1-carboxylate, benzyl 2-hydroxymethyl-2,3-dihydroindole-1-carboxylate, 1h-indole-1-carboxylicacid, 2,3-dihydro-2-hydroxymethyl-, phenylmethyl ester, acmc-20c3wg, 1-benzyloxycarbonylindoline-2-methanol, +/-1-benzyloxycarbonyl-2-hydroxymethylindoline
PubChem CID 2776402
IUPAC Name benzyl 2-(hydroxymethyl)-2,3-dihydroindole-1-carboxylate
SMILES C1C(N(C2=CC=CC=C21)C(=O)OCC3=CC=CC=C3)CO

Specifications

CAS Min % 94.5
CAS Max % 100.0
Assay Percent Range 95%
Packaging Amber Glass Bottle
Quantity 1g
Formula Weight 283.33
Percent Purity 95%
Chemical Name or Material Benzyl 2-(hydroxymethyl)-1-indolinecarboxylate