missing translation for 'onlineSavingsMsg'
Learn More

Benzyl 2-bromoacetate, 96%, Thermo Scientific™

Supplier:  Thermo Scientific Chemicals 278040010

Quantity 1kg

 View more versions of this product

Catalog No. AC278040010

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description

Description

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Chemical Identifiers

5437-45-6
229.08
JHVLLYQQQYIWKX-UHFFFAOYSA-N
62576
C1=CC=C(C=C1)COC(=O)CBr
C9H9BrO2
MFCD00000190
benzyl bromoacetate, bromoacetic acid benzyl ester, acetic acid, bromo-, phenylmethyl ester, merbac 35, caswell no. 082a, acetic acid, bromo-, benzyl ester, unii-64u2rk18d3, phenylmethyl 2-bromoacetate, epa pesticide chemical code 008710, benzyl bromacetate
benzyl 2-bromoacetate

Specifications

Benzyl 2-bromoacetate
5437-45-6
100.0
166.0°C to 170.0°C (22.0mmHg)
Authentic
Glass bottle
1.5430 to 1.5450
MFCD00000190
benzyl bromoacetate, bromoacetic acid benzyl ester, acetic acid, bromo-, phenylmethyl ester, merbac 35, caswell no. 082a, acetic acid, bromo-, benzyl ester, unii-64u2rk18d3, phenylmethyl 2-bromoacetate, epa pesticide chemical code 008710, benzyl bromacetate
JHVLLYQQQYIWKX-UHFFFAOYSA-N
benzyl 2-bromoacetate
62576
96%
96%
95.0
1.4400g/mL
>109°C
95% min. (GC)
C9H9BrO2
1kg
1.44
Solubility in water: insoluble.
C1=CC=C(C=C1)COC(=O)CBr
229.08
229.08
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

EINECSNumber : 226-611-4

RTECSNumber : AF5957215

SDS
Documents

Documents

Promotions

Promotions

Promo
Expires: 06-30-2027
Get Up to $1,800 in Chemicals with Purchase
Spend $250 or more on Thermo Fisher Scientific chemicals and get up to $1,800 to spend on additional chemicals.

Adding...