missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Benzyl 2,3-O-isopropylidene-alpha-D-mannofuranoside, Thermo Scientific™
Supplier: Thermo Scientific Chemicals J6625403
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
SolubilitySoluble in chloroform, dichloromethane or DMSO
Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
Specifications
| Benzyl 2,3-O-isopropylidene-alpha-D-mannofuranoside | |
| White | |
| MFCD02683277 | |
| benzyl 2,3-o-isopropylidene-alpha-d-mannofuranoside, benzyl 2,3-o-isopropylidene-, a-d-mannofuranoside, phenylmethyl 2,3-o-1-methylethylidene-, benzyl 3xi-2,3-o-1-methylethylidene-alpha-d-erythro-hexofuranoside, 1r-1-4r,6s,6ar-6-benzyloxy-2,2-dimethyl-tetrahydrofuro 3,4-d 1,3 dioxol-4-yl ethane-1,2-diol | |
| NKCNUNUEQRYLNZ-MLQJFRMTSA-N | |
| (1R)-1-[(3aR,4S,6R)-2,2-dimethyl-4-phenylmethoxy-3a,4,6,6a-tetrahydrofuro[3,4-d][1,3]dioxol-6-yl]ethane-1,2-diol | |
| 57370143 | |
| Powder |
| 20689-03-6 | |
| C16H22O6 | |
| 1 g | |
| Soluble in chloroform, dichloromethane or DMSO | |
| CC1(OC2C(O1)C(OC2C(CO)O)OCC3=CC=CC=C3)C | |
| 310.346 | |
| 310.34 |
Chemical Identifiers
| 20689-03-6 | |
| 310.346 | |
| NKCNUNUEQRYLNZ-MLQJFRMTSA-N | |
| 57370143 | |
| CC1(OC2C(O1)C(OC2C(CO)O)OCC3=CC=CC=C3)C |
| C16H22O6 | |
| MFCD02683277 | |
| benzyl 2,3-o-isopropylidene-alpha-d-mannofuranoside, benzyl 2,3-o-isopropylidene-, a-d-mannofuranoside, phenylmethyl 2,3-o-1-methylethylidene-, benzyl 3xi-2,3-o-1-methylethylidene-alpha-d-erythro-hexofuranoside, 1r-1-4r,6s,6ar-6-benzyloxy-2,2-dimethyl-tetrahydrofuro 3,4-d 1,3 dioxol-4-yl ethane-1,2-diol | |
| (1R)-1-[(3aR,4S,6R)-2,2-dimethyl-4-phenylmethoxy-3a,4,6,6a-tetrahydrofuro[3,4-d][1,3]dioxol-6-yl]ethane-1,2-diol |
Safety and Handling
TSCA : No
Recommended Storage : Store at -20°C