missing translation for 'onlineSavingsMsg'
Learn More

Benzydamine hydrochloride

Catalog No. AAJ6118006
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ6118006 5 g
AAJ6118014 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAJ6118006 Supplier Thermo Scientific Chemicals Supplier No. J6118006

CAS: 132-69-4 | C19H24ClN3O | 345.871 g/mol

Nonsteroidal analgesic and anti-inflammatory compound. A prostaglandin synthase inhibitor

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Nonsteroidal analgesic and anti-inflammatory compound
  • A prostaglandin synthase inhibitor

Chemical Identifiers

CAS 132-69-4
Molecular Formula C19H24ClN3O
Molecular Weight (g/mol) 345.871
MDL Number MFCD00078957
InChI Key HNNIWKQLJSNAEQ-UHFFFAOYSA-N
Synonym benzydamine hydrochloride, benzydamine hcl, benalgin, benzindamine hydrochloride, benzidamine hydrochloride, benzidan, benzyrin, difflam, dorinamin, enzamin
PubChem CID 65464
IUPAC Name 3-(1-benzylindazol-3-yl)oxy-N,N-dimethylpropan-1-amine;hydrochloride
SMILES CN(C)CCCOC1=NN(C2=CC=CC=C21)CC3=CC=CC=C3.Cl

Specifications

Melting Point 156°C to 161°C
Color White
Quantity 5 g
Merck Index 14,1122
Solubility Information Soluble in water,ethanol,chloroform,n-butanol or DMSO /n
Formula Weight 345.87
Physical Form Crystalline Powder
Chemical Name or Material Benzydamine hydrochloride
Hazard Category H302+H312+H332-H319
Hazard Statement GHS H Statement
H302-H312-H332-H319
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Causes serious eye irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P363-P501c
EINECSNumber 205-076-0
RTECSNumber NK7875000
TSCA No
Recommended Storage Keep cold

RUO – Research Use Only