missing translation for 'onlineSavingsMsg'
Learn More

Benzydamine Hydrochloride 97.0+%, TCI America™

Catalog No. B37515G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B37515G 5 g
B375125G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B37515G Supplier TCI America Supplier No. B37515G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 132-69-4
Molecular Formula C19H24ClN3O
Molecular Weight (g/mol) 345.871
MDL Number MFCD00078957
InChI Key HNNIWKQLJSNAEQ-UHFFFAOYSA-N
Synonym benzydamine hydrochloride, benzydamine hcl, benalgin, benzindamine hydrochloride, benzidamine hydrochloride, benzidan, benzyrin, difflam, dorinamin, enzamin
PubChem CID 65464
IUPAC Name 3-(1-benzylindazol-3-yl)oxy-N,N-dimethylpropan-1-amine;hydrochloride
SMILES CN(C)CCCOC1=NN(C2=CC=CC=C21)CC3=CC=CC=C3.Cl

Specifications

Melting Point 160°C
Color White
Quantity 5 g
Formula Weight 345.87
Percent Purity ≥97.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material Benzydamine Hydrochloride
RTECSNumber NK7875000
TSCA No
Recommended Storage Refrigerator