missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Benzoylurea 98.0+%, TCI America™
$55.84 - $546.75
Chemical Identifiers
| CAS | 614-22-2 |
|---|---|
| Molecular Formula | C8H8N2O2 |
| Molecular Weight (g/mol) | 164.16 |
| MDL Number | MFCD00025441 |
| InChI Key | HRYILSDLIGTCOP-UHFFFAOYSA-N |
| Synonym | benzoylurea, 1-benzoylurea, n-benzoylurea, urea, benzoyl, benzoic acid, ureide, n-aminocarbonyl benzamide, benzamide, n-aminocarbonyl, benzamide, n-aminocarbonyl-9ci, benzoyl urea, benzoylcarbamide |
| PubChem CID | 69190 |
| ChEBI | CHEBI:39416 |
| IUPAC Name | benzoylurea |
| SMILES | NC(=O)NC(=O)C1=CC=CC=C1 |
Chemical Identifiers
| 614-22-2 | |
| 164.16 | |
| HRYILSDLIGTCOP-UHFFFAOYSA-N | |
| 69190 | |
| benzoylurea |
| C8H8N2O2 | |
| MFCD00025441 | |
| benzoylurea, 1-benzoylurea, n-benzoylurea, urea, benzoyl, benzoic acid, ureide, n-aminocarbonyl benzamide, benzamide, n-aminocarbonyl, benzamide, n-aminocarbonyl-9ci, benzoyl urea, benzoylcarbamide | |
| CHEBI:39416 | |
| NC(=O)NC(=O)C1=CC=CC=C1 |
Specifications
| 614-22-2 | |
| White | |
| MFCD00025441 | |
| benzoylurea, 1-benzoylurea, n-benzoylurea, urea, benzoyl, benzoic acid, ureide, n-aminocarbonyl benzamide, benzamide, n-aminocarbonyl, benzamide, n-aminocarbonyl-9ci, benzoyl urea, benzoylcarbamide | |
| NC(=O)NC(=O)C1=CC=CC=C1 | |
| 164.16 | |
| CHEBI:39416 | |
| ≥98.0% (HPLC,N) | |
| Benzoylurea |
| 210°C | |
| C8H8N2O2 | |
| 25 g | |
| HRYILSDLIGTCOP-UHFFFAOYSA-N | |
| benzoylurea | |
| 69190 | |
| 164.16 | |
| Crystalline Powder |
Safety and Handling
RTECSNumber : YR9070000
TSCA : No