missing translation for 'onlineSavingsMsg'
Learn More

Benzoylurea 98.0+%, TCI America™

Catalog No. B040025G
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B040025G 25 g
B0400500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B040025G Supplier TCI America Supplier No. B040025G

Chemical Identifiers

CAS 614-22-2
Molecular Formula C8H8N2O2
Molecular Weight (g/mol) 164.16
MDL Number MFCD00025441
InChI Key HRYILSDLIGTCOP-UHFFFAOYSA-N
Synonym benzoylurea, 1-benzoylurea, n-benzoylurea, urea, benzoyl, benzoic acid, ureide, n-aminocarbonyl benzamide, benzamide, n-aminocarbonyl, benzamide, n-aminocarbonyl-9ci, benzoyl urea, benzoylcarbamide
PubChem CID 69190
ChEBI CHEBI:39416
IUPAC Name benzoylurea
SMILES NC(=O)NC(=O)C1=CC=CC=C1

Specifications

Melting Point 210°C
Color White
Quantity 25 g
Formula Weight 164.16
Percent Purity ≥98.0% (HPLC,N)
Physical Form Crystalline Powder
Chemical Name or Material Benzoylurea
RTECSNumber YR9070000
TSCA No