missing translation for 'onlineSavingsMsg'
Learn More

Benzoylhydrazine 98.0+%, TCI America™

Catalog No. B0205500G
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B0205500G 500 g
B020525G 25 g
B0205100G 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. B0205500G Supplier TCI America Supplier No. B0205500G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 613-94-5
Molecular Formula C7H8N2O
Molecular Weight (g/mol) 136.15
MDL Number MFCD00007596
InChI Key WARCRYXKINZHGQ-UHFFFAOYSA-N
Synonym benzoylhydrazine, benzhydrazide, benzoic hydrazide, benzoyl hydrazine, benzoic acid, hydrazide, benzoic acid hydrazide, benzohydrazine, benzoylhydrazide, benzoyl hydrazide, hydrazine, benzoyl
PubChem CID 11955
ChEBI CHEBI:38454
IUPAC Name benzohydrazide
SMILES NNC(=O)C1=CC=CC=C1

Specifications

Melting Point 114°C
Color White
Quantity 500 g
Formula Weight 136.15
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material Benzoylhydrazine
RTECSNumber DH1575000
TSCA Yes