missing translation for 'onlineSavingsMsg'
Learn More

Benzoylhydrazine, 98%

Catalog No. AC105775000
Change view
Click to view available options
Quantity:
100 g
500 g
Packaging:
Plastic bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC105775000 500 g Plastic bottle
AC105771000 100 g Plastic bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC105775000 Supplier Thermo Scientific Chemicals Supplier No. 105775000
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 613-94-5 | C7H8N2O | 136.15 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 613-94-5
Molecular Formula C7H8N2O
Molecular Weight (g/mol) 136.15
MDL Number MFCD00007596
InChI Key WARCRYXKINZHGQ-UHFFFAOYSA-N
Synonym benzoylhydrazine, benzhydrazide, benzoic hydrazide, benzoyl hydrazine, benzoic acid, hydrazide, benzoic acid hydrazide, benzohydrazine, benzoylhydrazide, benzoyl hydrazide, hydrazine, benzoyl
PubChem CID 11955
ChEBI CHEBI:38454
IUPAC Name benzohydrazide
SMILES NNC(=O)C1=CC=CC=C1

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 113.0°C to 117.0°C
Color White to Beige
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Plastic bottle
Linear Formula C6H5CONHNH2
Quantity 500 g
Beilstein 09,319
Solubility Information Solubility in water: soluble
Formula Weight 136.15
Percent Purity 98%
Physical Form Needle-like Crystals or Powder
Chemical Name or Material Benzoylhydrazine, 98%
Show More Show Less
Hazard Category GHS Signal Word: Warning
EINECSNumber 210-363-9
RTECSNumber DH1575000
TSCA TSCA

RUO – Research Use Only