missing translation for 'onlineSavingsMsg'
Learn More

Benzoyleneurea, 98%

Catalog No. p-7044685
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0776306 5 g
AAL0776314 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL0776306 Supplier Thermo Scientific Chemicals Supplier No. L0776306
Only null left

CAS: 86-96-4 | C8H6N2O2 | 162.148 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 86-96-4
Molecular Formula C8H6N2O2
Molecular Weight (g/mol) 162.148
MDL Number MFCD00006699
InChI Key SDQJTWBNWQABLE-UHFFFAOYSA-N
Synonym benzoyleneurea, quinazoline-2,4 1h,3h-dione, 2,4 1h,3h-quinazolinedione, quinazoline-2,4-diol, 2,4-dihydroxyquinazoline, quinazolinedione, benzouracil, quinazoline-2,4-dione, urea, benzoylene, 1,2,3,4-tetrahydroquinazoline-2,4-dione
PubChem CID 64048
IUPAC Name 1H-quinazoline-2,4-dione
SMILES C1=CC=C2C(=C1)C(=O)NC(=O)N2

Specifications

Melting Point >300°C
Quantity 5 g
Beilstein 383776
Formula Weight 162.15
Percent Purity 98%
Chemical Name or Material Benzoyleneurea
EINECSNumber 201-712-6
RTECSNumber VA1390000
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only