missing translation for 'onlineSavingsMsg'
Learn More

Benzoylcholine Iodide 98.0+%, TCI America™

Catalog No. B010825G
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B010825G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B010825G Supplier TCI America Supplier No. B010825G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 17518-43-3
Molecular Formula C12H18INO2
Molecular Weight (g/mol) 335.19
MDL Number MFCD00038726
InChI Key XILQCEGWGBSDSH-UHFFFAOYSA-M
Synonym benzoylcholine iodide, benzoylcholineiodide, 2-benzoyloxy-n,n,n-trimethylethanaminium iodide, 2-benzoyloxyethyl trimethylammonium iodide, ethanaminium, 2-benzoyloxy-n,n,n-trimethyl-, iodide, ethanaminium, 2-benzoyloxy-n,n,n-trimethyl-, iodide 1:1, 2-benzoyloxy ethyl trimethylazanium iodide, choline, iodide benzoate, choline, benzoate, acmc-1buu4
PubChem CID 87143
IUPAC Name [2-(benzoyloxy)ethyl]trimethylazanium iodide
SMILES [I-].C[N+](C)(C)CCOC(=O)C1=CC=CC=C1

Specifications

Color White-Yellow
Quantity 25 g
Formula Weight 335.19
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Benzoylcholine Iodide