missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Benzoylacetonitrile 98.0+%, TCI America™
Supplier: TCI America B009825G
Specifications
| Benzoylacetonitrile | |
| 81°C | |
| C9H7NO | |
| 25 g | |
| ZJRCIQAMTAINCB-UHFFFAOYSA-N | |
| 3-oxo-3-phenylpropanenitrile | |
| 64799 | |
| 145.16 | |
| Crystalline Powder |
| 614-16-4 | |
| 160°C | |
| MFCD00001942 | |
| benzoylacetonitrile, 2-cyanoacetophenone, phenacyl cyanide, 3-oxo-3-phenyl-propionitrile, cyanomethyl phenyl ketone, acetonitrile, benzoyl, cyanoacetophenone, 3-phenyl-3-ketopropionitrile, benzenepropanenitrile, beta-oxo, benzoyl acetonitrile | |
| O=C(CC#N)C1=CC=CC=C1 | |
| 145.16 | |
| CHEBI:51855 | |
| ≥98.0% (GC) |
Chemical Identifiers
| 614-16-4 | |
| 145.16 | |
| ZJRCIQAMTAINCB-UHFFFAOYSA-N | |
| 64799 | |
| 3-oxo-3-phenylpropanenitrile |
| C9H7NO | |
| MFCD00001942 | |
| benzoylacetonitrile, 2-cyanoacetophenone, phenacyl cyanide, 3-oxo-3-phenyl-propionitrile, cyanomethyl phenyl ketone, acetonitrile, benzoyl, cyanoacetophenone, 3-phenyl-3-ketopropionitrile, benzenepropanenitrile, beta-oxo, benzoyl acetonitrile | |
| CHEBI:51855 | |
| O=C(CC#N)C1=CC=CC=C1 |
Safety and Handling
TSCA : Yes