missing translation for 'onlineSavingsMsg'
Learn More

Benzoylacetonitrile, 98+%

Catalog No. AC402000050
Change view
Click to view available options
Quantity:
5 g
Packaging:
Glass bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC402000050 5 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC402000050 Supplier Thermo Scientific Chemicals Supplier No. 402000050

CAS: 614-16-4 | C9H7NO | 145.16 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 614-16-4
Molecular Formula C9H7NO
Molecular Weight (g/mol) 145.16
MDL Number MFCD00001942
InChI Key ZJRCIQAMTAINCB-UHFFFAOYSA-N
Synonym benzoylacetonitrile, 2-cyanoacetophenone, phenacyl cyanide, 3-oxo-3-phenyl-propionitrile, cyanomethyl phenyl ketone, acetonitrile, benzoyl, cyanoacetophenone, 3-phenyl-3-ketopropionitrile, benzenepropanenitrile, beta-oxo, benzoyl acetonitrile
PubChem CID 64799
ChEBI CHEBI:51855
IUPAC Name 3-oxo-3-phenylpropanenitrile
SMILES O=C(CC#N)C1=CC=CC=C1

Specifications

CAS Min % 98.0
CAS Max % 100.0
Melting Point 78.0°C to 82.0°C
Boiling Point 160.0°C (10.0 mmHg)
Assay Percent Range 98+%
Packaging Glass bottle
Linear Formula C6H5COCH2CN
Quantity 5 g
Beilstein 10, 680
Solubility Information Solubility in water: slightly soluble
Formula Weight 145.16
Percent Purity 98+%
Chemical Name or Material Benzoylacetonitrile, ≥98%
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Harmful if swallowed.
Causes skin irritation.
Harmful in contact with skin.
Harmful if inhaled.
May cause respiratory irritation.
Causes serious eye irritation.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor
EINECSNumber 210-369-1
TSCA TSCA

RUO – Research Use Only