missing translation for 'onlineSavingsMsg'
Learn More

Benzoyl Leuco Methylene Blue 96.0+%, TCI America™

Catalog No. p-7118377
Change view
Click to view available options
Quantity:
1 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B02071G 1 g
B020725G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B02071G Supplier TCI America Supplier No. B02071G
Only null left

Chemical Identifiers

CAS 1249-97-4
Molecular Formula C23H23N3OS
Molecular Weight (g/mol) 389.517
MDL Number MFCD00059153
InChI Key ZKURGBYDCVNWKH-UHFFFAOYSA-N
PubChem CID 94975
IUPAC Name [3,7-bis(dimethylamino)phenothiazin-10-yl]-phenylmethanone
SMILES CN(C)C1=CC2=C(C=C1)N(C3=C(S2)C=C(C=C3)N(C)C)C(=O)C4=CC=CC=C4

Specifications

Melting Point 198°C
Color Green-Yellow
Quantity 1 g
Formula Weight 389.52
Percent Purity ≥96.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material Benzoyl Leuco Methylene Blue
EINECSNumber (5)-0910
RTECSNumber SN5449000
TSCA Yes