missing translation for 'onlineSavingsMsg'
Learn More

Benzoyl isocyanate, 90%, Tech.

CAS: 4461-33-0 | C8H5NO2 | 147.13 g/mol

$63.54 - $225.87

Chemical Identifiers

CAS 4461-33-0
Molecular Formula C8H5NO2
Molecular Weight (g/mol) 147.13
MDL Number MFCD00002032
InChI Key LURYMYITPCOQAU-UHFFFAOYSA-N
Synonym benzoylisocyanate, methanone, 1-phenyl-1-isocyanato, oxophenylmethanisocyanate, benzoyl isocynate, phconco, acmc-20alpt, pubchem7575, isocyanato phenyl methanone, isocyanato-phenyl-methanone, 9ci
PubChem CID 78210
IUPAC Name benzoyl isocyanate
SMILES C1=CC=C(C=C1)C(=O)N=C=O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC314280010
SDS View Documents
Thermo Scientific Chemicals
314280010
1 g Glass bottle
Each for $63.54
Only null left
 
AC314280050
SDS View Documents
Thermo Scientific Chemicals
314280050
5 g Glass bottle
Each for $225.87
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

4461-33-0
147.13
LURYMYITPCOQAU-UHFFFAOYSA-N
78210
C1=CC=C(C=C1)C(=O)N=C=O
C8H5NO2
MFCD00002032
benzoylisocyanate, methanone, 1-phenyl-1-isocyanato, oxophenylmethanisocyanate, benzoyl isocynate, phconco, acmc-20alpt, pubchem7575, isocyanato phenyl methanone, isocyanato-phenyl-methanone, 9ci
benzoyl isocyanate

Specifications

4461-33-0
100.0
1.1700g/mL
72°C
88% min. (GC)
C8H5NO2
C6H5CONCO
1 g
benzoylisocyanate, methanone, 1-phenyl-1-isocyanato, oxophenylmethanisocyanate, benzoyl isocynate, phconco, acmc-20alpt, pubchem7575, isocyanato phenyl methanone, isocyanato-phenyl-methanone, 9ci
C1=CC=C(C=C1)C(=O)N=C=O
147.13
147.13
Technical
Benzoyl isocyanate, tech., 90%
88.0
White
94°C to 96°C (21.0 mmHg)
Authentic
Glass bottle
1.5504 to 1.5524
MFCD00002032
1.17
LURYMYITPCOQAU-UHFFFAOYSA-N
benzoyl isocyanate
78210
90%
Solid
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Harmful in contact with skin.
Causes skin irritation.
Harmful if inhaled.
Harmful if swallowed.
May cause respiratory irritation.
Causes serious eye irritation.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

EINECSNumber : 224-717-5

SDS
Documents

Documents

RUO – Research Use Only