missing translation for 'onlineSavingsMsg'
Learn More

Benzoyl isocyanate, 90%, Tech.

Catalog No. AC314280010
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC314280010 1 g Glass bottle
AC314280050 5 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC314280010 Supplier Thermo Scientific Chemicals Supplier No. 314280010
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 4461-33-0 | C8H5NO2 | 147.13 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 4461-33-0
Molecular Formula C8H5NO2
Molecular Weight (g/mol) 147.13
MDL Number MFCD00002032
InChI Key LURYMYITPCOQAU-UHFFFAOYSA-N
Synonym benzoylisocyanate, methanone, 1-phenyl-1-isocyanato, oxophenylmethanisocyanate, benzoyl isocynate, phconco, acmc-20alpt, pubchem7575, isocyanato phenyl methanone, isocyanato-phenyl-methanone, 9ci
PubChem CID 78210
IUPAC Name benzoyl isocyanate
SMILES C1=CC=C(C=C1)C(=O)N=C=O

Specifications

CAS Min % 88.0
CAS Max % 100.0
Color White
Density 1.1700g/mL
Boiling Point 94°C to 96°C (21.0 mmHg)
Flash Point 72°C
Infrared Spectrum Authentic
Assay Percent Range 88% min. (GC)
Packaging Glass bottle
Refractive Index 1.5504 to 1.5524
Linear Formula C6H5CONCO
Quantity 1 g
Specific Gravity 1.17
Formula Weight 147.13
Percent Purity 90%
Grade Technical
Physical Form Solid
Chemical Name or Material Benzoyl isocyanate, tech., 90%
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Harmful in contact with skin.
Causes skin irritation.
Harmful if inhaled.
Harmful if swallowed.
May cause respiratory irritation.
Causes serious eye irritation.
Precautionary Statement GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 224-717-5

RUO – Research Use Only